Assay Details
| Gene Symbol: S6K2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337766.1 | NP_001327295.1 | 5 - 6 | 1491 | 144 | |
| NM_001337767.1 | NP_001327294.1 | 5 - 6 | 1402 | 144 | ||
| NM_111706.4 | NP_187484.1 | 4 - 5 | 1357 | 144 | ||
| NM_180212.2 | NP_850543.1 | 4 - 5 | 1720 | 144 | ||
| GenBank mRNA | AF325094.1 | - | 3 - 4 | 1004 | 144 | |
| AK316712.1 | - | 4 - 5 | 1475 | 144 | ||
| BT026369.1 | - | 3 - 4 | 1004 | 144 | ||
More Information
Set Membership: |
|

S6K2
Genomic Map