Assay Details
| Gene Symbol: BBC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035754.1 | NP_001030831.1 | 3 - 4 | 323 | 127 | |
| NM_001084791.1 | NP_001078260.1 | 3 - 4 | 298 | 127 | ||
| NM_001339393.1 | NP_001327179.1 | 3 - 4 | 634 | 127 | ||
| NM_001339394.1 | NP_001327180.1 | 3 - 4 | 660 | 127 | ||
| NM_114760.4 | NP_190470.1 | 3 - 4 | 308 | 127 | ||
| NM_180341.3 | NP_850672.1 | 3 - 4 | 306 | 127 | ||
| GenBank mRNA | AF428384.1 | - | 3 - 4 | 284 | 127 | |
| AK317079.1 | - | 3 - 4 | 324 | 127 | ||
| AY050854.1 | - | 3 - 4 | 288 | 127 | ||
| AY065137.1 | - | 3 - 4 | 294 | 127 | ||
| AY079381.1 | - | 2 - 3 | 240 | 127 | ||
| AY081595.1 | - | 2 - 3 | 240 | 127 | ||
| X75162.1 | - | 263 | 127 | |||
More Information
Set Membership: |
|

BBC1
Genomic Map