Assay Details
| Gene Symbol: PPME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105663.5 | NP_177152.2 | 3 - 4 | 781 | 160 | |
| GenBank mRNA | BT002992.1 | - | 3 - 4 | 779 | 160 | |
More Information
Set Membership: |
|

PPME1
Genomic Map