Assay Details
| Gene Symbol: ARAC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106234.3 | NP_177712.1 | 1 - 2 | 418 | 120 | |
| GenBank mRNA | AY062800.1 | - | 1 - 2 | 360 | 120 | |
| AY081600.1 | - | 1 - 2 | 103 | 120 | ||
| U52350.1 | - | 1 - 2 | 298 | 120 | ||
More Information
Set Membership: |
|

ARAC5
Genomic Map