Assay Details
| Gene Symbol: MYB107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337450.1 | NP_001326399.1 | 2 - 3 | 1064 | 144 | |
| NM_111164.3 | NP_186944.1 | 3 - 4 | 1048 | 144 | ||
| GenBank mRNA | AF249310.2 | - | 3 - 4 | 1023 | 144 | |
More Information
Set Membership: |
|

MYB107
Genomic Map