Assay Details
| Gene Symbol: AT3G03870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111258.4 | NP_566215.1 | 1 - 2 | 1143 | 152 | |
| Gene Symbol: PYK10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK226844.1 | - | 2233 | 152 | ||
More Information
Set Membership: |
|

AT3G03870
Genomic Map