Assay Details
| Gene Symbol: TRX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103588.5 | NP_175128.1 | 1 - 2 | 253 | 126 | |
| GenBank mRNA | AF360227.1 | - | 1 - 2 | 155 | 126 | |
| AK118035.1 | - | 1 - 2 | 155 | 126 | ||
| AK221784.1 | - | 1 - 2 | 155 | 126 | ||
| AY040028.1 | - | 1 - 2 | 92 | 126 | ||
| AY087159.1 | - | 1 - 2 | 157 | 126 | ||
| Z35476.1 | - | 138 | 126 | |||
More Information
Set Membership: |
|

TRX5
Genomic Map