Assay Details
Gene Symbol: PAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103619.5 | NP_175158.1 | 1 - 2 | 453 | 177 | |
GenBank mRNA | AF043527.1 | - | 1 - 2 | 373 | 177 | |
AY057649.1 | - | 1 - 2 | 395 | 177 | ||
AY141996.1 | - | 1 - 2 | 352 | 177 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
