Assay Details
| Gene Symbol: AT1G52245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160947.2 | NP_001154419.1 | 2 - 3 | 548 | 137 | |
| GenBank mRNA | BT010683.1 | - | 2 - 3 | 359 | 137 | |
| BT012415.1 | - | 1 - 2 | 137 | 137 | ||
More Information
Set Membership: |
|

AT1G52245
Genomic Map