Assay Details
| Gene Symbol: PK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337768.1 | NP_001326081.1 | 1 - 2 | 1453 | 108 | |
| NM_111707.4 | NP_187485.1 | 2 - 3 | 936 | 108 | ||
| GenBank mRNA | AY065230.1 | - | 2 - 3 | 858 | 108 | |
| AY096555.1 | - | 1 - 2 | 604 | 108 | ||
| D42056.1 | - | 1 - 2 | 837 | 108 | ||
| L29030.1 | - | 2 - 3 | 860 | 108 | ||
More Information
Set Membership: |
|

PK1
Genomic Map