Assay Details
| Gene Symbol: PK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337768.1 | NP_001326081.1 | 3 - 4 | 1836 | 142 | |
| NM_111707.4 | NP_187485.1 | 4 - 5 | 1319 | 142 | ||
| GenBank mRNA | AY065230.1 | - | 4 - 5 | 1241 | 142 | |
| AY096555.1 | - | 3 - 4 | 987 | 142 | ||
| D42056.1 | - | 3 - 4 | 1220 | 142 | ||
| L29030.1 | - | 4 - 5 | 1243 | 142 | ||
More Information
Set Membership: |
|

PK1
Genomic Map