Assay Details
| Gene Symbol: PK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337768.1 | NP_001326081.1 | 4 - 5 | 1940 | 145 | |
| NM_111707.4 | NP_187485.1 | 5 - 6 | 1423 | 145 | ||
| GenBank mRNA | AY065230.1 | - | 5 - 6 | 1345 | 145 | |
| AY096555.1 | - | 4 - 5 | 1091 | 145 | ||
| D42056.1 | - | 4 - 5 | 1324 | 145 | ||
| L29030.1 | - | 5 - 6 | 1347 | 145 | ||
More Information
Set Membership: |
|

PK1
Genomic Map