Assay Details
| Gene Symbol: ACT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338359.1 | NP_001327851.1 | 2 - 3 | 1494 | 148 | |
| NM_112764.4 | NP_188508.1 | 3 - 4 | 1454 | 148 | ||
| GenBank mRNA | AF428330.1 | - | 3 - 4 | 1242 | 148 | |
| AK230311.1 | - | 1831 | 148 | |||
| AK317305.1 | - | 3 - 4 | 1247 | 148 | ||
| AY064043.1 | - | 3 - 4 | 1247 | 148 | ||
| AY087751.1 | - | 3 - 4 | 1252 | 148 | ||
| AY096381.1 | - | 2 - 3 | 1072 | 148 | ||
More Information
Set Membership: |
|

ACT2
Genomic Map