Assay Details
| Gene Symbol: SK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113078.1 | NP_566692.1 | 1 | 340 | 124 | |
| GenBank mRNA | DQ056601.1 | - | 1 | 340 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SK8| Gene Symbol: SK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113078.1 | NP_566692.1 | 1 | 340 | 124 | |
| GenBank mRNA | DQ056601.1 | - | 1 | 340 | 124 | |
Set Membership: |
|