Assay Details
| Gene Symbol: SK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113080.2 | NP_566694.1 | 1 | 400 | 136 | |
| GenBank mRNA | DQ446683.1 | - | 1 | 334 | 136 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SK9| Gene Symbol: SK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113080.2 | NP_566694.1 | 1 | 400 | 136 | |
| GenBank mRNA | DQ446683.1 | - | 1 | 334 | 136 | |
Set Membership: |
|