Assay Details
| Gene Symbol: MYB0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113708.2 | NP_189430.1 | 1 - 2 | 239 | 135 | |
| GenBank mRNA | AF495524.1 | - | 1 - 2 | 138 | 135 | |
| AY519590.1 | - | 1 - 2 | 138 | 135 | ||
More Information
Set Membership: |
|

MYB0
Genomic Map