Assay Details
| Gene Symbol: AT5G11880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121226.5 | NP_568252.1 | 4 - 5 | 1277 | 129 | |
| GenBank mRNA | AY049266.1 | - | 4 - 5 | 1015 | 129 | |
| AY142042.1 | - | 4 - 5 | 991 | 129 | ||
More Information
Set Membership: |
|

AT5G11880
Genomic Map