Assay Details
| Gene Symbol: bHLH38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115556.4 | NP_191256.1 | 1 - 2 | 462 | 94 | |
| GenBank mRNA | AF488576.1 | - | 1 - 2 | 311 | 94 | |
| AK221542.1 | - | 1 - 2 | 367 | 94 | ||
| BT026419.1 | - | 1 - 2 | 311 | 94 | ||
More Information
Set Membership: |
|

bHLH38
Genomic Map