Assay Details
| Gene Symbol: SDR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203222.1 | NP_001190151.1 | 2 - 3 | 366 | 132 | |
| NM_001340081.1 | NP_001326283.1 | 1 - 2 | 514 | 132 | ||
| NM_115986.4 | NP_191681.1 | 2 - 3 | 449 | 132 | ||
| GenBank mRNA | AY045884.1 | - | 2 - 3 | 311 | 132 | |
| AY091305.1 | - | 2 - 3 | 273 | 132 | ||
More Information
Set Membership: |
|

SDR1
Genomic Map