Assay Details
| Gene Symbol: SDR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203222.1 | NP_001190151.1 | 4 - 5 | 594 | 112 | |
| NM_001340081.1 | NP_001326283.1 | 3 - 4 | 742 | 112 | ||
| NM_115986.4 | NP_191681.1 | 4 - 5 | 677 | 112 | ||
| GenBank mRNA | AY045884.1 | - | 4 - 5 | 539 | 112 | |
| AY091305.1 | - | 4 - 5 | 501 | 112 | ||
More Information
Set Membership: |
|

SDR1
Genomic Map