Assay Details
| Gene Symbol: BRCA2(IV) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340223.1 | NP_001328738.1 | 17 - 18 | 3712 | 111 | |
| NM_001340224.1 | NP_001328737.1 | 18 - 19 | 3637 | 111 | ||
| NM_116219.5 | NP_191913.3 | 19 - 20 | 3531 | 111 | ||
| GenBank mRNA | AJ488304.1 | - | 19 - 20 | 3391 | 111 | |
More Information
Set Membership: |
|

BRCA2(IV)
Genomic Map