Assay Details
| Gene Symbol: AT4G08780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116948.2 | NP_192618.1 | 3 - 4 | 613 | 150 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G08780| Gene Symbol: AT4G08780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116948.2 | NP_192618.1 | 3 - 4 | 613 | 150 | |
Set Membership: |
|