Assay Details
| Gene Symbol: RPS18C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117048.4 | NP_192718.1 | 3 - 4 | 460 | 158 | |
| GenBank mRNA | AF386941.1 | - | 3 - 4 | 447 | 158 | |
| AY086334.1 | - | 3 - 4 | 457 | 158 | ||
| BT006539.1 | - | 2 - 3 | 354 | 158 | ||
More Information
Set Membership: |
|

RPS18C
Genomic Map