Assay Details
| Gene Symbol: AT4G13263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117398.3 | NP_567402.1 | 1 | 162 | 143 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G13263| Gene Symbol: AT4G13263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117398.3 | NP_567402.1 | 1 | 162 | 143 | |
Set Membership: |
|