Assay Details
| Gene Symbol: AT4G15620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117653.6 | NP_193297.1 | 1 - 2 | 521 | 158 | |
| GenBank mRNA | BT002974.1 | - | 1 - 2 | 332 | 158 | |
| BT004455.1 | - | 1 - 2 | 239 | 158 | ||
More Information
Set Membership: |
|

AT4G15620
Genomic Map