Assay Details
| Gene Symbol: AT4G15630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117654.3 | NP_567473.1 | 1 - 2 | 506 | 118 | |
| GenBank mRNA | AK229320.1 | - | 1 - 2 | 331 | 118 | |
| BT028921.1 | - | 1 - 2 | 239 | 118 | ||
More Information
Set Membership: |
|

AT4G15630
Genomic Map