Assay Details
| Gene Symbol: NLM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341319.1 | NP_001328744.1 | 2 - 3 | 498 | 129 | |
| NM_001341320.1 | NP_001328743.1 | 2 - 3 | 477 | 129 | ||
| NM_118021.4 | NP_567572.1 | 2 - 3 | 498 | 129 | ||
| GenBank mRNA | AY063796.1 | - | 2 - 3 | 477 | 129 | |
| AY117197.1 | - | 2 - 3 | 399 | 129 | ||
More Information
Set Membership: |
|

NLM1
Genomic Map