Assay Details
| Gene Symbol: AT4G27130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118847.4 | NP_194443.1 | 3 - 4 | 516 | 168 | |
| GenBank mRNA | AY037234.1 | - | 3 - 4 | 448 | 168 | |
| AY088288.1 | - | 3 - 4 | 496 | 168 | ||
More Information
Set Membership: |
|

AT4G27130
Genomic Map