Assay Details
| Gene Symbol: CSLC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119308.3 | NP_194887.1 | 2 - 3 | 1678 | 124 | |
| GenBank mRNA | BT003334.1 | - | 2 - 3 | 1282 | 124 | |
| BT008423.1 | - | 2 - 3 | 1027 | 124 | ||
| KJ138731.1 | - | 2 - 3 | 1027 | 124 | ||
More Information
Set Membership: |
|

CSLC5
Genomic Map