Assay Details
| Gene Symbol: CSLC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119308.3 | NP_194887.1 | 4 - 5 | 2079 | 150 | |
| GenBank mRNA | BT003334.1 | - | 4 - 5 | 1683 | 150 | |
| BT008423.1 | - | 4 - 5 | 1428 | 150 | ||
| KJ138731.1 | - | 4 - 5 | 1428 | 150 | ||
More Information
Set Membership: |
|

CSLC5
Genomic Map