Assay Details
| Gene Symbol: GCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120644.3 | NP_196181.1 | 1 - 2 | 239 | 129 | |
| GenBank mRNA | BT030394.1 | - | 1 - 2 | 129 | 129 | |
More Information
Set Membership: |
|

GCP2
Genomic Map