Assay Details
| Gene Symbol: MYB29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120851.2 | NP_196386.1 | 1 - 2 | 552 | 155 | |
| GenBank mRNA | AY035145.1 | - | 1 - 2 | 457 | 155 | |
| AY059078.1 | - | 1 - 2 | 131 | 155 | ||
| AY519617.1 | - | 1 - 2 | 131 | 155 | ||
More Information
Set Membership: |
|

MYB29
Genomic Map