Assay Details
| Gene Symbol: AT5G08670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120953.4 | NP_568203.2 | 7 - 8 | 1341 | 146 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G08670| Gene Symbol: AT5G08670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120953.4 | NP_568203.2 | 7 - 8 | 1341 | 146 | |
Set Membership: |
|