Assay Details
Gene Symbol: AT5G38030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123162.4 | NP_198619.1 | 4 - 5 | 1246 | 151 | |
GenBank mRNA | AF372972.1 | - | 4 - 5 | 1049 | 151 | |
BT002707.1 | - | 4 - 5 | 982 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
