Assay Details
| Gene Symbol: VIM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123317.4 | NP_198771.1 | 3 - 4 | 1260 | 133 | |
| GenBank mRNA | AK221256.1 | - | 3 - 4 | 1238 | 133 | |
| BT010573.1 | - | 3 - 4 | 1085 | 133 | ||
More Information
Set Membership: |
|

VIM3
Genomic Map