Assay Details
| Gene Symbol: VIM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123317.4 | NP_198771.1 | 8 - 9 | 1868 | 154 | |
| GenBank mRNA | AK221256.1 | - | 8 - 9 | 1846 | 154 | |
| BT010573.1 | - | 8 - 9 | 1693 | 154 | ||
More Information
Set Membership: |
|

VIM3
Genomic Map