Assay Details
| Gene Symbol: GSTT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344393.1 | NP_001330832.1 | 3 - 4 | 560 | 97 | |
| NM_123487.4 | NP_198938.1 | 4 - 5 | 445 | 97 | ||
More Information
Set Membership: |
|

GSTT3
Genomic Map