Assay Details
| Gene Symbol: GSTT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344393.1 | NP_001330832.1 | 6 - 7 | 1033 | 131 | |
| NM_001344394.1 | NP_001330831.1 | 6 - 7 | 936 | 131 | ||
| NM_123487.4 | NP_198938.1 | 7 - 8 | 918 | 131 | ||
More Information
Set Membership: |
|

GSTT3
Genomic Map