Assay Details
| Gene Symbol: GSTT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123489.4 | NP_198940.3 | 4 - 5 | 311 | 129 | |
| GenBank mRNA | AY139996.1 | - | 4 - 5 | 304 | 129 | |
| BT008361.1 | - | 4 - 5 | 257 | 129 | ||
More Information
Set Membership: |
|

GSTT2
Genomic Map