Assay Details
| Gene Symbol: MYB96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125641.4 | NP_201053.2 | 1 - 2 | 412 | 123 | |
| GenBank mRNA | AK228977.1 | - | 1 - 2 | 359 | 123 | |
| BT024888.1 | - | 1 - 2 | 129 | 123 | ||
More Information
Set Membership: |
|

MYB96
Genomic Map