Assay Details
| Gene Symbol: AT2G41980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129762.6 | NP_181729.1 | 4 - 5 | 1223 | 165 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G41980| Gene Symbol: AT2G41980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129762.6 | NP_181729.1 | 4 - 5 | 1223 | 165 | |
Set Membership: |
|