Assay Details
| Gene Symbol: TPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130040.3 | NP_566026.1 | 3 - 4 | 391 | 158 | |
| GenBank mRNA | AY042898.1 | - | 3 - 4 | 391 | 158 | |
| AY081493.1 | - | 3 - 4 | 307 | 158 | ||
More Information
Set Membership: |
|

TPK2
Genomic Map