Assay Details
| Gene Symbol: SLK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148375.6 | NP_680741.5 | 8 - 9 | 1261 | 179 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLK3| Gene Symbol: SLK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148375.6 | NP_680741.5 | 8 - 9 | 1261 | 179 | |
Set Membership: |
|