Assay Details
| Gene Symbol: SCL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100627.5 | NP_172233.1 | 2 | 2607 | 85 | |
| GenBank mRNA | AK316793.1 | - | 2 | 2497 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCL14| Gene Symbol: SCL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100627.5 | NP_172233.1 | 2 | 2607 | 85 | |
| GenBank mRNA | AK316793.1 | - | 2 | 2497 | 85 | |
Set Membership: |
|