Assay Details
| Gene Symbol: SARA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100788.3 | NP_172390.1 | 3 | 671 | 82 | |
| GenBank mRNA | AY090921.1 | - | 3 | 547 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SARA1A| Gene Symbol: SARA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100788.3 | NP_172390.1 | 3 | 671 | 82 | |
| GenBank mRNA | AY090921.1 | - | 3 | 547 | 82 | |
Set Membership: |
|