Assay Details
| Gene Symbol: HB31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101311.6 | NP_172896.1 | 2 | 1712 | 78 | |
| NM_202097.1 | NP_973826.1 | 2 | 1071 | 78 | ||
| GenBank mRNA | AB493456.1 | - | 1 | 897 | 78 | |
| AY074266.1 | - | 2 | 1061 | 78 | ||
| AY096738.1 | - | 1 | 897 | 78 | ||
More Information
Set Membership: |
|

HB31
Genomic Map