Assay Details
| Gene Symbol: PUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102920.3 | NP_174466.1 | 2 | 2029 | 93 | |
| NM_202218.3 | NP_973947.1 | 2 | 1962 | 93 | ||
| GenBank mRNA | AK317082.1 | - | 2 | 1817 | 93 | |
More Information
Set Membership: |
|

PUT2
Genomic Map