Assay Details
| Gene Symbol: BAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114827.4 | NP_190536.1 | 2 | 3335 | 75 | |
| GenBank mRNA | AK228595.1 | - | 2 | 3232 | 75 | |
| BT005722.1 | - | 2 | 3232 | 75 | ||
More Information
Set Membership: |
|

BAM2
Genomic Map