Assay Details
| Gene Symbol: AT1G53030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104181.5 | NP_175711.1 | 2 | 180 | 62 | |
| GenBank mRNA | AF349685.1 | - | 2 | 121 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G53030| Gene Symbol: AT1G53030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104181.5 | NP_175711.1 | 2 | 180 | 62 | |
| GenBank mRNA | AF349685.1 | - | 2 | 121 | 62 | |
Set Membership: |
|