Assay Details
| Gene Symbol: KIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121602.4 | NP_197101.1 | 3 | 444 | 152 | |
| GenBank mRNA | AY072302.1 | - | 3 | 252 | 152 | |
| X55053.1 | - | 246 | 152 | |||
More Information
Set Membership: |
|

KIN2
Genomic Map